Learn More
2-(Bromomethyl)-5-(trifluoromethyl)furan, 97%, Thermo Scientific™
£86.35 - £246.00
Chemical Identifiers
CAS | 17515-77-4 |
---|---|
Molecular Formula | C6H4BrF3O |
Molecular Weight (g/mol) | 229.00 |
MDL Number | MFCD03086219 |
InChI Key | YNHVBNGRNNVEMD-UHFFFAOYSA-N |
Synonym | 2-bromomethyl-5-trifluoromethyl furan, 2-bromomethyl-5-trifluoromethyl-furan, furan, 2-bromomethyl-5-trifluoromethyl, pubchem13345, 5-trifluoromethyl furfuryl bromide, 2-bromomethyl-5-trifloromethyl furan, 5-bromomethyl-2-trifluoromethyl furan |
PubChem CID | 2794605 |
IUPAC Name | 2-(bromomethyl)-5-(trifluoromethyl)furan |
SMILES | FC(F)(F)C1=CC=C(CBr)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10520742
|
Thermo Scientific Maybridge
MO00141CB |
250 mg |
£86.35
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10242502
|
Thermo Scientific Maybridge
MO00141DA |
1 g |
£109.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10200275
|
Thermo Scientific Maybridge
MO00141DE |
5 g |
£246.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
17515-77-4 | |
229.00 | |
YNHVBNGRNNVEMD-UHFFFAOYSA-N | |
2794605 | |
FC(F)(F)C1=CC=C(CBr)O1 |
C6H4BrF3O | |
MFCD03086219 | |
2-bromomethyl-5-trifluoromethyl furan, 2-bromomethyl-5-trifluoromethyl-furan, furan, 2-bromomethyl-5-trifluoromethyl, pubchem13345, 5-trifluoromethyl furfuryl bromide, 2-bromomethyl-5-trifloromethyl furan, 5-bromomethyl-2-trifluoromethyl furan | |
2-(bromomethyl)-5-(trifluoromethyl)furan |
Specifications
17515-77-4 | |
97% | |
C6H4BrF3O | |
2-bromomethyl-5-trifluoromethyl furan, 2-bromomethyl-5-trifluoromethyl-furan, furan, 2-bromomethyl-5-trifluoromethyl, pubchem13345, 5-trifluoromethyl furfuryl bromide, 2-bromomethyl-5-trifloromethyl furan, 5-bromomethyl-2-trifluoromethyl furan | |
FC(F)(F)C1=CC=C(CBr)O1 | |
229.00 | |
228.99 | |
2-(Bromomethyl)-5-(trifluoromethyl)furan |
97.0 | |
Amber Glass Bottle | |
MFCD03086219 | |
YNHVBNGRNNVEMD-UHFFFAOYSA-N | |
2-(bromomethyl)-5-(trifluoromethyl)furan | |
2794605 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger