missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-5-(trifluoromethyl)furan, 97%, Thermo Scientific™

Product Code. 10200275
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10200275 5 g 5g
10520742 250 mg 250mg
10242502 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10200275 Supplier Thermo Scientific Maybridge Supplier No. MO00141DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17515-77-4
Molecular Formula C6H4BrF3O
Molecular Weight (g/mol) 229.00
MDL Number MFCD03086219
InChI Key YNHVBNGRNNVEMD-UHFFFAOYSA-N
Synonym 2-bromomethyl-5-trifluoromethyl furan, 2-bromomethyl-5-trifluoromethyl-furan, furan, 2-bromomethyl-5-trifluoromethyl, pubchem13345, 5-trifluoromethyl furfuryl bromide, 2-bromomethyl-5-trifloromethyl furan, 5-bromomethyl-2-trifluoromethyl furan
PubChem CID 2794605
IUPAC Name 2-(bromomethyl)-5-(trifluoromethyl)furan
SMILES FC(F)(F)C1=CC=C(CBr)O1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 228.99
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)-5-(trifluoromethyl)furan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.