missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol, 99%, Thermo Scientific Chemicals

Product Code. 10628031
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19184-65-7
Molecular Formula C5H11BrO3
Molecular Weight (g/mol) 199.05
MDL Number MFCD00135306
InChI Key MMHHBAUIJVTLFZ-UHFFFAOYSA-N
Synonym 2-bromomethyl-2-hydroxymethyl propane-1,3-diol, 2-bromomethyl-2-hydroxymethyl-1,3-propanediol, 1,3-propanediol, 2-bromomethyl-2-hydroxymethyl, tris hydroxymethyl bromomethyl methane, monobromoneopentyltriol, 2-bromoethylidynetrimethanol, pentaerythritol monobromohydrin, 1, 2-bromomethyl-2-hydroxymethyl, 2,2-bis hydroxymethyl-3-bromopropan-1-ol, 2,2-bis hydroxymethyl-3-hydroxypropyl bromide
PubChem CID 87954
IUPAC Name 2-(bromomethyl)-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CBr)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 77°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C(CH2OH)3
Quantity 5 g
Formula Weight 199.05
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Bromomethyl)-2-(hydroxymethyl)-1, 3-propanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'