missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-1,3-dioxolane, 97%

Product Code. 11489566
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489566 500 g 500g
11469566 25 g 25g
11479566 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489566 Supplier Thermo Scientific Alfa Aesar Supplier No. A19312.36
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4360-63-8 | C4H7BrO2 | 167.002 g/mol

2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4360-63-8
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.002
MDL Number MFCD00003214
InChI Key CKIIJIDEWWXQEA-UHFFFAOYSA-N
Synonym 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
PubChem CID 78068
IUPAC Name 2-(bromomethyl)-1,3-dioxolane
SMILES C1COC(O1)CBr

Specifications

Density 1.628
Boiling Point 175°C
Flash Point 62°C (145°F)
Refractive Index 1.482
Quantity 500 g
Beilstein 103309
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material 2-Bromomethyl-1,3-dioxolane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.