missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomethyl-1,3-dioxolane, 97%

Product Code. 10562611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10562611 100 mL Glass bottle 100mL
10164500 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10562611 Supplier Thermo Scientific Acros Supplier No. 209841000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4360-63-8 | C4H7BrO2 | 167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4360-63-8
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167
MDL Number MFCD00003214
InChI Key CKIIJIDEWWXQEA-UHFFFAOYSA-N
PubChem CID 78068
IUPAC Name 2-(bromomethyl)-1,3-dioxolane
SMILES C1COC(O1)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.6100g/mL
Boiling Point 80°C to 82°C (27.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4810 to 1.483
Quantity 100 mL
Beilstein 19, II, 8
Specific Gravity 1.61
Synonym 2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
Formula Weight 167
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromomethyl-1, 3-dioxolane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromomethyl-1,3-dioxolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.