missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-1,3-benzothiazole, 95%, Thermo Scientific™

Product Code. 10431474
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 106086-78-6
Molecular Formula C8H6BrNS
Molecular Weight (g/mol) 228.107
InChI Key WFLCAOGKZQTOIG-UHFFFAOYSA-N
Synonym 2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole
PubChem CID 2776258
IUPAC Name 2-(bromomethyl)-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)CBr

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 228.11
Percent Purity 95%
Chemical Name or Material 2-(Bromomethyl)-1, 3-benzothiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'