missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomesitylene, 99%

Product Code. 11399659
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
Unit Size:
100g
250g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399659 100 g 100g
11484993 50 g 50g
11494993 250 g 250g
11309669 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11399659 Supplier Thermo Scientific Alfa Aesar Supplier No. A12277.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 576-83-0 | C9H11Br | 199.091 g/mol

2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.

Solubility
Insoluble in water(20°C) .

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, excess heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-83-0
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00000073
InChI Key RRTLQRYOJOSPEA-UHFFFAOYSA-N
Synonym 2-bromomesitylene, 2,4,6-trimethylbromobenzene, mesityl bromide, bromomesitylene, benzene, 2-bromo-1,3,5-trimethyl, 2,4,6-trimethybromombenzene, mesitylene, 2-bromo, 2,4,6-trimethylphenyl bromide, 1-bromo-2,4,6-trimethylbenzene, 2-bromo-1,3,5-trimethyl-benzene
PubChem CID 68473
IUPAC Name 2-bromo-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)Br)C

Specifications

Melting Point 2°C
Density 1.322
Boiling Point 224°C to 226°C
Flash Point 96°C (204°F)
Refractive Index 1.552
Quantity 100 g
Beilstein 1907245
Sensitivity Light sensitive
Solubility Information Insoluble in water(20°C) .
Formula Weight 199.1
Percent Purity 99%
Chemical Name or Material 2-Bromomesitylene
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromomesitylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromomesitylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.