missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomalonaldehyde, 97+%

Product Code. 10336704 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336704 5 g Glass bottle 5g
10123404 1 g Glass bottle 1g
10575924 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10336704 Supplier Thermo Scientific Acros Supplier No. 397900050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2065-75-0 | C3H3BrO2 | 150.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2065-75-0
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.95
InChI Key SURMYNZXHKLDFO-UHFFFAOYSA-N
Synonym 2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
PubChem CID 74945
IUPAC Name 2-bromopropanedial
SMILES C(=O)C(C=O)Br

Specifications

Melting Point 132.0°C to 136.0°C
Assay Percent Range 98.5 % min. (Acid)
Packaging Glass bottle
Linear Formula BrCH(CHO)2
Quantity 5 g
Formula Weight 150.95
Percent Purity 97+%
Chemical Name or Material 2-Bromomalonaldehyde
compliance-icons compliance-icons
Product Identifier
  • 2-Bromomalonaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.