missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisovaleric acid, 96%

Product Code. 11459642
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459642 10 g 10g
11469642 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459642 Supplier Thermo Scientific Alfa Aesar Supplier No. A10756.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 565-74-2 | C5H9BrO2 | 181.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-74-2
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.029
MDL Number MFCD00004210
InChI Key UEBARDWJXBGYEJ-UHFFFAOYSA-N
Synonym 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb
PubChem CID 11268
IUPAC Name 2-bromo-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)Br

Specifications

Melting Point 36°C to 42°C
Boiling Point 124°C to 126°C (20 mmHg)
Odor Acrid
Linear Formula (CH3)2CHCHBrCO2H
Quantity 10 g
UN Number UN3261
Merck Index 14,1422
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material 2-Bromoisovaleric acid
compliance-icons compliance-icons
Product Identifier
  • 2-Bromoisovaleric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromoisovaleric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.