missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisobutyric Acid, TRC

Product Code. 30346885
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346885 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346885 Supplier Toronto Research Chemicals Supplier No. TRCB684645100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoisobutyric Acid
CAS 2052-01-9
Molecular Formula C4H7BrO2
Quantity 100 g
Synonym 2-Bromo-2-methyl-propanoic Acid, α-Bromo-isobutyric Acid, 2-Bromo-2-methyl-propionic Acid, 2-Bromo-2-methylpropanoic Acid, 2-Bromo-2-methylpropionic Acid, 2-Bromoisobutyric Acid, Bromoisobutyric Acid, Isobromobutyric Acid, NSC 41213, α-Bromo-α-methylpropionic Acid, α-Bromoisobutyric Acid
InChI Formula InChI=1S/C6H11BrO2/c1-4-9-5(8)6(2,3)7/h4H2,1-3H3
SMILES CC(C)(Br)C(=O)O
IUPAC Name ethyl 2-bromo-2-methylpropanoate
Molecular Weight (g/mol) 167
Formula Weight 165.9629
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.