missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisobutyric acid, 98%

Product Code. 11452548
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11452548

Brand: Thermo Scientific Alfa Aesar A15854.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2052-01-9 | C4H7BrO2 | 167.00 g/mol

2-Bromoisobutyric acid is used in the synthesis of dextran macroinitiator for atom transfer radical polymerization (ATRP) by partial esterification of hydroxyl group of the polysaccharide. The multifunctional silica colloids were prepared by coating them with 2-bromo-2-methylpropionic acid stabilized quantum dots.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoisobutyric acid is used in the synthesis of dextran macroinitiator for atom transfer radical polymerization (ATRP) by partial esterification of hydroxyl group of the polysaccharide. The multifunctional silica colloids were prepared by coating them with 2-bromo-2-methylpropionic acid stabilized quantum dots.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-01-9
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00004168
InChI Key XXSPGBOGLXKMDU-UHFFFAOYSA-N
Synonym 2-bromoisobutyric acid, 2-bromo-2-methylpropionic acid, isobromobutyric acid, propanoic acid, 2-bromo-2-methyl, alpha-bromoisobutyric acid, .alpha.-bromoisobutyric acid, 2-bromo-2-methyl-propanoic acid, propionic acid, 2-bromo-2-methyl, unii-821p2r6b0a, ccris 9046
PubChem CID 74924
IUPAC Name 2-bromo-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)Br

Specifications

Melting Point 42°C to 46°C
Boiling Point 198°C to 200°C
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN3261
Beilstein 1745543
Merck Index 14,1421
Solubility Information Soluble in alcohol and ether. Slightly soluble in water.
Formula Weight 167
Percent Purity 98%
Chemical Name or Material 2-Bromoisobutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromoisobutyric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Bromoisobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.