missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoindene, 98%

Product Code. 11451050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451050 5 g 5g
11441050 1 g 1g
11461050 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451050 Supplier Thermo Scientific Alfa Aesar Supplier No. B25474.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10485-09-3 | C9H7Br | 195.059 g/mol

2-Bromoindene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoindene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10485-09-3
Molecular Formula C9H7Br
Molecular Weight (g/mol) 195.059
MDL Number MFCD06797863
InChI Key CCUYEVNCRQDQRF-UHFFFAOYSA-N
Synonym 2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate
PubChem CID 575586
IUPAC Name 2-bromo-1H-indene
SMILES C1C2=CC=CC=C2C=C1Br

Specifications

Melting Point 35°C to 37°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 195.06
Percent Purity 98%
Chemical Name or Material 2-Bromoindene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.