missing translation for 'onlineSavingsMsg'
Learn More

2-Bromohexane, 98%, Thermo Scientific™

Product Code. 11332116
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332116 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11332116 Supplier Thermo Scientific Alfa Aesar Supplier No. L03364.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromohexane can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Store away from strong bases. Incompatible with oxidizing agents and bases. Light-sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3377-86-4
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.074
MDL Number MFCD00039183
InChI Key NEBYCXAKZCQWAW-UHFFFAOYSA-N
Synonym hexane, 2-bromo, 2-hexyl bromide, dl, 2-bromo-hexane, acmc-209t5g, nebycxakzcqwaw-uhfffaoysa
PubChem CID 18805
IUPAC Name 2-bromohexane
SMILES CCCCC(C)Br

Specifications

Boiling Point 146°C to 147°C
Flash Point 47°C (116°F)
Odor Pleasant
Quantity 5 g
UN Number UN1993
Beilstein 1731328
Formula Weight 165.08
Percent Purity 98%
Chemical Name or Material 2-Bromohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.