missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofuran, 97%, Thermo Scientific™

Product Code. 10306514
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306514 5 g Amber Glass Bottle 5g
10572822 1 g Amber Glass Bottle 1g
10517464 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306514 Supplier Thermo Scientific Maybridge Supplier No. CC02010DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 584-12-3 , 7440-50-8 , 110-00-9 , 144-55-8
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00216609
InChI Key OYMCMWPHMPODNK-UHFFFAOYSA-N
Synonym furan, 2-bromo, bromofuran, 2-bromo-furan, 2-furyl bromide, alpha-furyl bromide, 2-bromofurane, 5-bromofuran, pubchem6929, acmc-209m4d
PubChem CID 2776190
IUPAC Name 2-bromofuran
SMILES BrC1=CC=CO1

Specifications

CAS Min % 96.5, 0.05, 0.45, 0.35
CAS Max % 98.5, 0.15, 0.55, 0.45
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 146.97
Percent Purity 97%
Chemical Name or Material 2-Bromofuran, stabilized with Copper (0.1%), Furan (0.5%) and Sodium Bicarbonate (0.4%)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.