missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorene, 95%

Product Code. 11341967
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11341967 25 g 25g
11331967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341967 Supplier Thermo Scientific Alfa Aesar Supplier No. L07589.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1133-80-8 | C13H9Br | 245.119 g/mol

2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromofluorene acts as a nonintrusive end-capping reagent in order to control molecular weights as well as in the generation of well-defined oligomers. It is used to prepare 2-bromo-9-cyclohepta-2,4,6-trienyl-9H-fluorene and 9,9-dihexylflorene. In addition to this, it is used in the preparation of poly(di-n-hexylfluorene)s end capped with 2-bromofluorene, 2-bromo-9,9-di-n-hexylfluorene and 9-bromoanthracene through Ni(0) mediated polymerization.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1133-80-8
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001115
InChI Key FXSCJZNMWILAJO-UHFFFAOYSA-N
Synonym 2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
PubChem CID 14336
IUPAC Name 2-bromo-9H-fluorene
SMILES C1C2=CC=CC=C2C3=C1C=C(C=C3)Br

Specifications

Melting Point 110°C to 114°C
Boiling Point 185°C (135 mmHg)
Quantity 25 g
Beilstein 1946701
Solubility Information Soluble in chloroform and ethyl acetate.
Formula Weight 245.12
Percent Purity 95%
Chemical Name or Material 2-Bromofluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.