missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl acetate, 97%

Product Code. 10048139 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10048139 25 g Glass bottle 25g
10646270 5 g Glass bottle 5g
10493340 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10048139 Supplier Thermo Scientific Acros Supplier No. 160090250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 927-68-4 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-68-4
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000235
InChI Key RGHQKFQZGLKBCF-UHFFFAOYSA-N
Synonym bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb
PubChem CID 13564
IUPAC Name 2-bromoethyl acetate
SMILES CC(=O)OCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -13.8°C
Color Colorless to Yellow
Density 1.5100g/mL
Boiling Point 160.0°C to 162.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2Br
Refractive Index 1.4547
Quantity 25 g
Specific Gravity 1.51
Formula Weight 167
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromoethyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.