missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethylamine Hydrobromide, TRC

Product Code. 30334490
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334490 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334490 Supplier Toronto Research Chemicals Supplier No. TRCB683750250G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoethylamine Hydrobromide
CAS 2576-47-8
Molecular Formula C2H7Br2N
Quantity 250 g
Synonym 1-Amino-2-bromoethane Hydrobromide, 2-Aminoethyl Bromide Hydrobromide, 2-Bromoethanamine Hydrobromide, 2-Bromoethylammonium Bromide, beta-Bromoethylamine Hydrobromide
InChI Formula InChI=1S/C2H6BrN.BrH/c3-1-2-4;/h1-2,4H2;1H
SMILES Br.NCCBr
IUPAC Name 2-bromoethanamine;hydrobromide
Molecular Weight (g/mol) 204.89
Formula Weight 202.8945
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.