missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethylamine Hydrobromide, >98.0%, MP Biomedicals™

Product Code. 11499021 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499021 25 g 25g
11409031 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499021 Supplier MP Biomedicals Supplier No. 0215487525
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2576-47-8
Molecular Formula C2H7Br2N
Molecular Weight (g/mol) 204.893
InChI Key WJAXXWSZNSFVNG-UHFFFAOYSA-N
Synonym 2-bromoethylamine hydrobromide, 2-bromoethanamine hydrobromide, 2-aminoethyl bromide hydrobromide, 2-bromoethylamine hbr, 2-bromoethylammonium bromide, ethanamine, 2-bromo-, hydrobromide, 2-bromoethan-1-amine hydrobromide, bromoethylamine hydrobromide, aminoethyl bromide hbr, ethylamine, hydrobromide
PubChem CID 2774217
IUPAC Name 2-bromoethanamine;hydrobromide
SMILES C(CBr)N.Br

Specifications

Color White
Density 1.581g/cm3
Assay Percent Range >98%
Quantity 25 g
Formula Weight 204.9
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoethylamine Hydrobromide, Quality Control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.