missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)triphenylphosphonium bromide, 98%

Product Code. 11436606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436606 5 g 5g
11446606 25 g 25g
11456606 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436606 Supplier Thermo Scientific Alfa Aesar Supplier No. B24833.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7301-93-1 | C20H19Br2P | 450.15 g/mol

(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7301-93-1
Molecular Formula C20H19Br2P
Molecular Weight (g/mol) 450.15
MDL Number MFCD04039352
InChI Key ZHLVWYZWQPNQDQ-UHFFFAOYSA-M
Synonym 2-bromoethyl triphenylphosphonium bromide, phosphonium, 2-bromoethyl triphenyl-, bromide, 2-bromoethyl triphenylphosohonium bromide, 2-bromoethyl triphenylphosphanium bromide, 2-bromoethyl triphenyl phosphanium bromide, 2-bromoethyl triphenyl phosphonium bromide, phosphonium, 2-bromoethyl triphenyl-,bromide
PubChem CID 22210408
IUPAC Name 2-bromoethyl(triphenyl)phosphanium;bromide
SMILES [Br-].BrCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 186°C to 189°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 450.15
Percent Purity 98%
Chemical Name or Material (2-Bromoethyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.