missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromoethyl ethyl ether, 95%, stab. with 0.5% Potassium carbonate

Product Code. 11316766
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316766 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11316766 Supplier Alfa Aesar™ Supplier No. L04034.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
2-Bromoethyl ethyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Specifications

CAS 592-55-2
Boiling Point 127°C to 128°C
Flash Point 21°C (69°F)
Molecular Formula C4H9BrO
Linear Formula BrCH2CH2OCH2CH3
MDL Number MFCD00000237
Quantity 25 g
UN Number UN2340
Beilstein 1731362
Synonym 2-bromoethyl ethyl ether, 2-ethoxyethyl bromide, 2-bromoethoxyethane, ethane, 1-bromo-2-ethoxy, 2-bromodiethyl ether, 1-ethoxy-2-bromoethane, 1-bromo-2-ethoxy-ethane, ether, 2-bromoethyl ethyl, 2-bromoethylethylether, 1-bromo-3-oxapentane
InChI Key MMYKTRPLXXWLBC-UHFFFAOYSA-N
SMILES CCOCCBr
IUPAC Name 1-bromo-2-ethoxyethane
Molecular Weight (g/mol) 153.02
PubChem CID 61141
Formula Weight 153.02
Percent Purity 95%
Chemical Name or Material 2-Bromoethyl ethyl ether, stabilized with 0.5% Potassium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.