missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)cyclohexane, 99%

Product Code. 11397436
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397436 25 g 25g
11377436 1 g 1g
11387436 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397436 Supplier Thermo Scientific Alfa Aesar Supplier No. L09466.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1647-26-3 | C8H15Br | 191.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1647-26-3
Molecular Formula C8H15Br
Molecular Weight (g/mol) 191.11
MDL Number MFCD00019398
InChI Key JRQAAYVLPPGEHT-UHFFFAOYSA-N
Synonym 2-bromoethyl cyclohexane, 2-cyclohexylethyl bromide, cyclohexane, 2-bromoethyl, 2-cyclohexylethylbromide, 1-bromo-2-cyclohexylethane, cyclohexylethylbromide, bromoethyl cyclohexane, cyclohexylethyl bromide, acmc-1bzct, 1-bromo-2cyclohexylethane
PubChem CID 15440
IUPAC Name 2-bromoethylcyclohexane
SMILES BrCCC1CCCCC1

Specifications

Density 1.221
Boiling Point 70°C to 71°C (6 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.49
Quantity 25 g
Beilstein 1919816
Formula Weight 191.12
Percent Purity 99%
Chemical Name or Material (2-Bromoethyl)cyclohexane
compliance-icons
Product Identifier
  • (2-Bromoethyl)cyclohexane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.