missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl acrylate, 94%, stab. with 900-1500ppm 4-methoxyphenol

Product Code. 11353196
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353196 5 g 5g
11343196 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353196 Supplier Thermo Scientific Alfa Aesar Supplier No. L12502.06
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4823-47-6 | C5H7BrO2 | 179.013 g/mol

2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.

Solubility
Miscible with water.

Notes
Light and heat sensitive Incompatible with strong oxidizing agents, polymerizing agents and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4823-47-6
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.013
MDL Number MFCD00013541
InChI Key CDZAAIHWZYWBSS-UHFFFAOYSA-N
PubChem CID 20963
IUPAC Name 2-bromoethyl prop-2-enoate
SMILES C=CC(=O)OCCBr

Specifications

Boiling Point 52°C to 53°C (5 mmHg)
Quantity 5 g
Beilstein 1749713
Synonym 2-bromoethyl acrylate, acrylic acid, 2-bromoethyl ester, 2-propenoic acid, 2-bromoethyl ester, ccris 4753, acrylic acid 2-bromoethyl ester, 2-bromoethylacrylate, 2-bromoethyl=acrylate, acmc-20ao1p, acrylic acid 2-bromo-ethyl ester, 2-propenoic acid,2-bromoethyl ester
Solubility Information Miscible with water.
Formula Weight 179.01
Percent Purity 94%
Chemical Name or Material 2-Bromoethyl acrylate, stabilized with 0.1% 4-methoxyphenol
compliance-icons
Product Identifier
  • 2-Bromoethyl acrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromoethyl acrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.