missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanol, TRC

Product Code. 30343777
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343777 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343777 Supplier Toronto Research Chemicals Supplier No. TRCB68473510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromoethanol
CAS 540-51-2
Molecular Formula C2 H5 Br O
Quantity 10 g
Synonym 2-Bromoethanol, 1-Bromo-2-ethanol, 1-Bromo-2-hydroxyethane, 2-Bromoethyl alcohol, 2-Hydroxyethyl bromide, Ethylene bromohydrin, Glycol bromohydrin, NSC 2869, β-Bromoethanol
InChI Formula InChI=1S/C2H5BrO/c3-1-2-4/h4H,1-2H2
SMILES OCCBr
IUPAC Name 2-bromoethanol
Molecular Weight (g/mol) 124.96
Formula Weight 123.9524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.