missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoethanesulfonic Acid Sodium Salt, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 2-Bromoethanesulfonic Acid Sodium Salt |
| CAS | 4263-52-9 |
| Molecular Formula | C2H4BrNaO3S |
| Quantity | 25 g |
| Synonym | Sodium 2-Bromoethanesulfonate, Sodium Bromoethylsulfonate, 2-Bromoethanesulfonic Acid Sodium Salt |
| InChI Formula | InChI=1S/C2H5BrO3S.Na/c3-1-2-7(4,5)6;/h1-2H2,(H,4,5,6);/q;+1/p-1 |
| SMILES | [Na+].[O-]S(=O)(=O)CCBr |
| IUPAC Name | sodium;2-bromoethanesulfonate |
| Molecular Weight (g/mol) | 211.01 |
| Formula Weight | 209.8962 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction