missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromodibenzofuran, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromodibenzofuran |
| CAS | 86-76-0 |
| Molecular Formula | C12 H7 Br O |
| Quantity | 2.5 g |
| Synonym | Dibenzofuran, 2-bromo-, 2-Bromodibenzofuran, 2-Bromodibenzo[b,d]furan, NSC 1735, 2-Monobromodibenzofuran, 2-MonoBDF, 2-MBDF |
| InChI Formula | InChI=1S/C12H7BrO/c13-8-5-6-12-10(7-8)9-3-1-2-4-11(9)14-12/h1-7H |
| SMILES | Brc1ccc2oc3ccccc3c2c1 |
| IUPAC Name | 2-bromodibenzofuran |
| Molecular Weight (g/mol) | 247.09 |
| Formula Weight | 245.968 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction