missing translation for 'onlineSavingsMsg'
Learn More

2-Bromodibenzofuran, 98%, Thermo Scientific Chemicals

Product Code. 15491869
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.

Solubility
Soluble in hot methanol.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-76-0
Molecular Formula C12H7BrO
Molecular Weight (g/mol) 247.091
MDL Number MFCD00092338
InChI Key CRJISNQTZDMKQD-UHFFFAOYSA-N
Synonym 2-bromodibenzo b,d furan, 2-bromo-dibenzofuran, dibenzofuran, 2-bromo, dibenzofuran, 2-bromo-8ci 9ci, 2-bromodibenzo b furan, 4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene
PubChem CID 6856
IUPAC Name 2-bromodibenzofuran
SMILES C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br

Specifications

Melting Point 108°C to 111°C
Quantity 5 g
Beilstein 142199
Solubility Information Soluble in hot methanol.
Formula Weight 247.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromodibenzofuran

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'