missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromocinnamic Acid, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromocinnamic Acid |
| CAS | 7499-56-1 |
| Molecular Formula | C9 H7 Br O2 |
| Quantity | 5 g |
| Synonym | o-Bromo-trans-cinnamic acid, 3-(2-Bromophenyl)acrylic acid |
| InChI Formula | InChI=1S/C9H7BrO2/c10-8-4-2-1-3-7(8)5-6-9(11)12/h1-6H,(H,11,12)/b6-5+ |
| SMILES | OC(=O)\C=C\c1ccccc1Br |
| IUPAC Name | (E)-3-(2-bromophenyl)prop-2-enoic acid |
| Molecular Weight (g/mol) | 227.05 |
| Formula Weight | 225.9629 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction