missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutane, 98%, Thermo Scientific Chemicals

Catalog Number p-7022344
Click to view available options
Quantity:
250 g
1000 g
78-76-2
C4H9Br
137.02
MFCD00000156
UPSXAPQYNGXVBF-UHFFFAOYSA-N
sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
6554
2-bromobutane
CCC(C)Br
78-76-2
C4H9Br
137.02
MFCD00000156
UPSXAPQYNGXVBF-UHFFFAOYSA-N
sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2
6554
2-bromobutane
CCC(C)Br

2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobutane is utilized in the synthesis of non-nucleosides. It is used as a solvent as well as an intermediate in organic synthesis. It is an important raw material used to prepare Grignard reagent by reacting with magnesium. Further, it reacts with hydrocyanic acid to prepare sodium salt to get (+-)-sec-butyl cyanide. It is an important raw material used to prepare Grignard reagent by reacting with magnesium.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, sodium, sodium oxides and potassium.
TRUSTED_SUSTAINABILITY
Melting Point °C to 112°C
Density 1.260
Boiling Point 91°C to 92°C
Flash Point 13°C (55°F)
Assay Percent Range 98%
Linear Formula CH3CHBrCH2CH3
Refractive Index 1.4369
Quantity 250 g
UN Number UN2339
Beilstein 505949
Merck Index 14,1554
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 137.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromobutane
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'