missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobiphenyl, 98%

Product Code. 11474887
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474887 25 g 25g
11454887 1 g 1g
11464887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474887 Supplier Thermo Scientific Alfa Aesar Supplier No. B21840.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2052-07-5 | C12H9Br | 233.11 g/mol

It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-07-5
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000065
InChI Key KTADSLDAUJLZGL-UHFFFAOYSA-N
Synonym 2-bromobiphenyl, o-bromobiphenyl, 2-bromo-biphenyl, 1,1'-biphenyl, 2-bromo, 2-bromo-1,1'-biphenyl, biphenyl, 2-bromo, 2-bromo-diphenyl, bromobiphenyl, 2-bromodiphenyl, 1,1'-biphenyl, bromo
PubChem CID 16329
IUPAC Name 1-bromo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2Br

Specifications

Melting Point 0°C to 2°C
Density 1.376
Boiling Point 298°C
Flash Point >110°C (230°F)
Refractive Index 1.629
Quantity 25 g
Beilstein 1863156
Solubility Information Insoluble in water.
Formula Weight 233.12
Percent Purity 98%
Chemical Name or Material 2-Bromobiphenyl
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.