missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzylamine, 98%, Thermo Scientific™
£20.77 - £199.70
Chemical Identifiers
CAS | 3959-05-5 |
---|---|
Molecular Formula | C7H8BrN |
Molecular Weight (g/mol) | 186.05 |
MDL Number | MFCD00025572 |
InChI Key | NOYASZMZIBFFNZ-UHFFFAOYSA-N |
Synonym | 2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo |
PubChem CID | 334072 |
IUPAC Name | (2-bromophenyl)methanamine |
SMILES | C1=CC=C(C(=C1)CN)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10038821
|
Thermo Scientific Acros
350510050 |
5g | Glass bottle |
£49.00
5g |
Estimated Shipment: 14-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11985241
|
Thermo Scientific Acros
350510250 |
25g | Glass bottle |
£199.70
25g |
Estimated Shipment: 14-07-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10282892
|
Thermo Scientific Acros
350510010 |
1g | Glass bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3959-05-5 | |
186.05 | |
NOYASZMZIBFFNZ-UHFFFAOYSA-N | |
334072 | |
C1=CC=C(C(=C1)CN)Br |
C7H8BrN | |
MFCD00025572 | |
2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo | |
(2-bromophenyl)methanamine |
Specifications
2-Bromobenzylamine | |
3959-05-5 | |
100.0 | |
97.5 % min. (GC) | |
C7H8BrN | |
MFCD00025572 | |
Glass bottle | |
2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo | |
NOYASZMZIBFFNZ-UHFFFAOYSA-N | |
(2-bromophenyl)methanamine | |
334072 | |
Liquid |
98% | |
97.5 | |
Undesignated | |
Authentic | |
BrC6H4CH2NH2 | |
12,III,2348 | |
1.5875 to 1.5910 | |
5g | |
C1=CC=C(C(=C1)CN)Br | |
186.05 | |
186.05 | |
98% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Bromobenzylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only