missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine, 96%, Thermo Scientific Chemicals

Product Code. 11459593
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmaceutical intermediates. As intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical intermediates. As intermediate in organic synthesis.

Solubility
Insoluble in Water. Soluble in Alcohol and Ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide..
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3959-05-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00025572
InChI Key NOYASZMZIBFFNZ-UHFFFAOYSA-N
Synonym 2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
PubChem CID 334072
IUPAC Name (2-bromophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Br

Specifications

Boiling Point 118°C to 119°C (9 mmHg)
Odor Amine-like
Quantity 25 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in Water. Soluble in Alcohol and Ether.
Formula Weight 186.06
Percent Purity 98%
Chemical Name or Material 2-Bromobenzylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'