missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl bromide, 98%

Product Code. 11443654
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443654 250 g 250g
11423654 10 g 10g
11433654 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443654 Supplier Thermo Scientific Alfa Aesar Supplier No. A11285.30
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3433-80-5 | C7H6Br2 | 249.933 g/mol

2-Bromobenzyl bromide was used in the synthesis of substituted quinazolines and 1,2,3,4-tetrahydroquinazolines, 2- and 3-substituted indenes, tris-2-bromotribenzylamine3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzyl bromide was used in the synthesis of substituted quinazolines and 1,2,3,4-tetrahydroquinazolines, 2- and 3-substituted indenes, tris-2-bromotribenzylamine3.

Solubility
Insoluble in water.

Notes
Store away from strong bases and oxidizing agents. Incompatible with amines, alcohols, steel.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 3433-80-5
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000173
InChI Key LZSYGJNFCREHMD-UHFFFAOYSA-N
Synonym 2-bromobenzyl bromide, o-bromobenzyl bromide, 1-bromo-2-bromomethyl benzene, 2-bromobenzylbromide, alpha,2-dibromotoluene, benzene, 1-bromo-2-bromomethyl, alpha,o-dibromotoluene, benzene, bromo bromomethyl, alpha-2-dibromotoluene, a,o-dibromotoluene
PubChem CID 76965
IUPAC Name 1-bromo-2-(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)Br

Spécification

Melting Point 29°C to 32°C
Boiling Point 127°C to 129°C (19 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
UN Number UN1759
Beilstein 971015
Formula Weight 249.94
Percent Purity 98%
Chemical Name or Material 2-Bromobenzyl bromide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.