missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl bromide, 98%

Product Code. 11423654
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423654 10 g 10g
11433654 50 g 50g
11443654 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423654 Supplier Thermo Scientific Alfa Aesar Supplier No. A11285.09
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3433-80-5 | C7H6Br2 | 249.933 g/mol

2-Bromobenzyl bromide was used in the synthesis of substituted quinazolines and 1,2,3,4-tetrahydroquinazolines, 2- and 3-substituted indenes, tris-2-bromotribenzylamine3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzyl bromide was used in the synthesis of substituted quinazolines and 1,2,3,4-tetrahydroquinazolines, 2- and 3-substituted indenes, tris-2-bromotribenzylamine3.

Solubility
Insoluble in water.

Notes
Store away from strong bases and oxidizing agents. Incompatible with amines, alcohols, steel.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3433-80-5
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000173
InChI Key LZSYGJNFCREHMD-UHFFFAOYSA-N
Synonym 2-bromobenzyl bromide, o-bromobenzyl bromide, 1-bromo-2-bromomethyl benzene, 2-bromobenzylbromide, alpha,2-dibromotoluene, benzene, 1-bromo-2-bromomethyl, alpha,o-dibromotoluene, benzene, bromo bromomethyl, alpha-2-dibromotoluene, a,o-dibromotoluene
PubChem CID 76965
IUPAC Name 1-bromo-2-(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)Br

Specifications

Melting Point 29°C to 32°C
Boiling Point 127°C to 129°C (19 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.619
Quantity 10 g
UN Number UN1759
Beilstein 971015
Solubility Information Insoluble in water.
Formula Weight 249.94
Percent Purity 98%
Chemical Name or Material 2-Bromobenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.