missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl bromide, 98%

Product Code. 10497190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10497190 100 g Glass bottle 100g
10153770 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10497190 Supplier Thermo Scientific Acros Supplier No. 181791000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3433-80-5 | C7H6Br2 | 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3433-80-5
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00000173
InChI Key LZSYGJNFCREHMD-UHFFFAOYSA-N
Synonym 2-bromobenzyl bromide, o-bromobenzyl bromide, 1-bromo-2-bromomethyl benzene, 2-bromobenzylbromide, alpha,2-dibromotoluene, benzene, 1-bromo-2-bromomethyl, alpha,o-dibromotoluene, benzene, bromo bromomethyl, alpha-2-dibromotoluene, a,o-dibromotoluene
PubChem CID 76965
IUPAC Name 1-bromo-2-(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 32.0°C
Color Colorless to Yellow
Boiling Point 129.0°C (19.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2Br
Refractive Index 1.6170 to 1.6190
Quantity 100 g
Beilstein 05, 308
Formula Weight 249.93
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 2-Bromobenzyl bromide, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.