Learn More
Thermo Scientific Chemicals Eriochrome Black T
Description
Eriochrome black T is used as an indicator in analytical chemistry. It is also used in the detection of rare earth metals. It finds application in dyeing and printing of wool fabric, dyeing of silk and nylon as well as in leather dyeing. It can be used in the spectrophotometric determination of diphenhydramine hydrochloride in pharmaceutical preparations. As a complexometric indicator, it is useful in the determination of hardness of water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Eriochrome black T is used as an indicator in analytical chemistry. It is also used in the detection of rare earth metals. It finds application in dyeing and printing of wool fabric, dyeing of silk and nylon as well as in leather dyeing. It can be used in the spectrophotometric determination of diphenhydramine hydrochloride in pharmaceutical preparations. As a complexometric indicator, it is useful in the determination of hardness of water.
Solubility
Soluble in water. Slightly soluble in alcohol and acetone.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
Specifications
Specifications
| Beilstein | 4121162 |
| Merck Index | 14,3667 |
| Solubility Information | Soluble in water. Slightly soluble in alcohol and acetone. |
| Quantity | 500 g |
| IUPAC Name | sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate |
| Formula Weight | 461.39 |
| Chemical Name or Material | Eriochrome Black T |
Safety and Handling
- Eriochrome Black T
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: 3-hydroxy-4-[(1-hydroxy-2-naphthalenyl)azo]-7-nitro-1-Naphthalenesulfonic acid monosodium salt
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.