missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl Alcohol, TRC

Product Code. 30336181
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize':
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30336181 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30336181 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB6799955G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromobenzyl Alcohol
CAS 18982-54-2
Molecular Formula C7H7BrO
Quantity 5 g
Synonym 2-Bromobenzenemethanol, 1-Bromo-2-(hydroxymethyl)benzene, 2-(Hydroxymethyl)phenyl Bromide, 2-Bromobenzyl Alcohol, o-Bromo-benzyl Alcohol, 2-Bromophenylmethanol, 2-Hydroxymethyl-1-bromobenzene, o-Bromobenzyl Alcohol
InChI Formula InChI=1S/C7H7BrO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5H2
SMILES OCc1ccccc1Br
IUPAC Name (2-bromophenyl)methanol
Molecular Weight (g/mol) 187.03
Formula Weight 185.968
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.