missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl alcohol, 98%

Product Code. 11417667
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417667 10 g 10g
11427667 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417667 Supplier Thermo Scientific Alfa Aesar Supplier No. A15700.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18982-54-2 | C7H7BrO | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18982-54-2
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00004600
InChI Key IOWGHQGLUMEZKG-UHFFFAOYSA-N
Synonym 2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746
PubChem CID 72850
IUPAC Name (2-bromophenyl)methanol
SMILES OCC1=CC=CC=C1Br

Specifications

Melting Point 78°C to 82°C
Quantity 10 g
Beilstein 2243418
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 2-Bromobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.