missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzotrifluoride, 98%

Product Code. 11479392
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479392 500 g 500g
11459392 25 g 25g
11469392 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479392 Supplier Thermo Scientific Alfa Aesar Supplier No. A10134.36
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 392-83-6 | C7H4BrF3 | 225.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-83-6
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000373
InChI Key RWXUNIMBRXGNEP-UHFFFAOYSA-N
Synonym 2-bromobenzotrifluoride, 1-bromo-2-trifluoromethyl benzene, o-bromobenzotrifluoride, benzene, 1-bromo-2-trifluoromethyl, 2-bromo-alpha,alpha,alpha-trifluorotoluene, 2-brombenzotrifluorid, o-bromobenzyltrifluoride, o-trifluoromethyl bromobenzene, 2-bromotrifluorotoluene, 2-brombenzotrifluorid czech
PubChem CID 9806
IUPAC Name 1-bromo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Br

Specifications

Density 1.662
Boiling Point 167°C to 168°C
Flash Point 68°C (154°F)
Odor Aromatic
Refractive Index 1.482
Quantity 500 g
Beilstein 1945750
Formula Weight 225.01
Percent Purity 99%
Chemical Name or Material 2-Bromobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.