missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoic Acid, TRC

Product Code. 30342374
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342374 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342374 Supplier Toronto Research Chemicals Supplier No. TRCB680500100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromobenzoic Acid
CAS 88-65-3
Molecular Formula C7H5BrO2
Quantity 100 g
Synonym o-Bromobenzoic Acid, NSC 176122, NSC 6976
InChI Formula InChI=1S/C7H5BrO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)
SMILES OC(=O)c1ccccc1Br
IUPAC Name 2-bromobenzoic acid
Molecular Weight (g/mol) 201.02
Formula Weight 199.9473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.