missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoic acid, 98%

Product Code. 11460164
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11460164

Brand: Thermo Scientific Alfa Aesar A13608.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-65-3 | C7H5BrO2 | 201.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-65-3
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00002402
InChI Key XRXMNWGCKISMOH-UHFFFAOYSA-N
Synonym o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657
PubChem CID 6940
IUPAC Name 2-bromobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)Br

Specifications

Melting Point 146°C to 150°C
Density 1.93
Quantity 250 g
Beilstein 971266
Formula Weight 201.02
Percent Purity 98%
Chemical Name or Material 2-Bromobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.