missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzimidazole, 99%

Product Code. 11346579
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346579 1 g 1g
11356579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346579 Supplier Thermo Scientific Alfa Aesar Supplier No. H26183.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 54624-57-6 | C7H5BrN2 | 197.035 g/mol

Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54624-57-6
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD00160009
InChI Key PHPYXVIHDRDPDI-UHFFFAOYSA-N
PubChem CID 2776281
IUPAC Name 2-bromo-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)Br

Specifications

Melting Point 195°C to 201°C
Quantity 1 g
Synonym 2-bromobenzimidazole, 2-bromo-1h-benzo d imidazole, 2-bromo-1h-1,3-benzodiazole, 2-bromo-1h-benzoimidazole, 1h-benzimidazole, 2-bromo, bromobenzimidazole, bromo-benzimidazole, pubchem16113, acmc-1an7o, 1h-benzimidazole,2-bromo
Solubility Information Insoluble in water.
Formula Weight 197.03
Percent Purity 99%
Chemical Name or Material 2-Bromobenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.