missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzhydrazide, 98+%

Product Code. 11334374
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11334374

Brand: Thermo Scientific Alfa Aesar L00937.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29418-67-5 | C7H7BrN2O | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29418-67-5
Molecular Formula C7H7BrN2O
Molecular Weight (g/mol) 215.05
MDL Number MFCD00051577
InChI Key PQNLAYLOCZKPIY-UHFFFAOYSA-N
Synonym 2-bromobenzhydrazide, 2-bromobenzoic hydrazide, benzoylhydrazine, o-bromo, 2-bromobenzoyl hydrazine, 2-bromo-benzoic acid hydrazide, 2-bromobenzenecarbohydrazide, 2-bromobenzoic acid hydrazide, bromobenzhydrazide, pubchem3742, 2-bromobenzoylhydrazine
PubChem CID 101460
IUPAC Name 2-bromobenzohydrazide
SMILES NNC(=O)C1=CC=CC=C1Br

Specifications

Melting Point 152°C to 154°C
Quantity 5 g
Beilstein 1941021
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 2-Bromobenzhydrazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.