missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzenesulfonyl chloride, 97%

Product Code. 10401501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10401501 5 g Glass bottle 5g
10606403 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10401501 Supplier Thermo Scientific Acros Supplier No. 306440050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-25-1 | C6H4BrClO2S | 255.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-25-1
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00051971
InChI Key VFPWGZNNRSQPBT-UHFFFAOYSA-N
Synonym 2-bromobenzene-1-sulfonyl chloride, 2-bromobenzenesulphonyl chloride, 2-bromosulfonyl chloride, o-bromobenzenesulfonyl chloride, 2-bromophenylsulfonyl chloride, 2-bromobenzenesulfonylchloride, 2-bromobenzenesulphonylchloride, 2-bromophenyl chlorosulfone, 2-bromo-benzenesulfonyl chloride, 2-bromobenzene sulfonyl chloride
PubChem CID 520403
IUPAC Name 2-bromobenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50.0°C to 51.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula BrC6H4SO2Cl
Quantity 5 g
Solubility Information Solubility in water: decomposes
Formula Weight 255.52
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzenesulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.