Learn More
2-Bromobenzamide, 98%
Description
2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromobenzamide has been used in microwave assisted one-pot synthesis of substituted 3-(phenylmethylene)isoindolin-1-ones, palladium-catalyzed synthesis of phenanthridinones and in the synthesis of new water-soluble iminophosphorane ligand.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 160°C to 162°C |
| Quantity | 25 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 200.03 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Bromobenzamide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.