missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzaldehyde ethylene acetal, 98+%, Thermo Scientific Chemicals

Product Code. p-7047674
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11365385

Brand: Thermo Scientific Alfa Aesar L19604.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34824-58-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00155124
InChI Key IWSGKSUCFVOWQU-UHFFFAOYSA-N
Synonym 2-2-bromophenyl-1,3-dioxolane, 2-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-2-bromophenyl, 1-bromo-2-1,3-dioxolan-2-yl benzene, 2-bromophenyldioxolane, 2-bromo benzaldehyde ethylene glycol acetal, 2-2-bromo-phenyl-1,3 dioxolane, pubchem3729, acmc-1csyy, bromobenzaldehyde ethylene acetal
PubChem CID 553534
IUPAC Name 2-(2-bromophenyl)-1,3-dioxolane
SMILES BrC1=CC=CC=C1C1OCCO1

Specifications

Boiling Point 126°C to 127°C (5 mmHg)
Refractive Index 1.568
Quantity 25 g
Beilstein 1284374
Solubility Information Difficult to mix in water.
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material 2-Bromobenzaldehyde ethylene acetal

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.