missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzaldehyde, 98%

Product Code. 11447347
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447347 25 g 25g
11457347 100 g 100g
11467347 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447347 Supplier Thermo Scientific Alfa Aesar Supplier No. A15065.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6630-33-7 | C7H5BrO | 185.02 g/mol

2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.

Solubility
Soluble in alcohol and benzene. Insoluble in water.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6630-33-7
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003300
InChI Key NDOPHXWIAZIXPR-UHFFFAOYSA-N
Synonym o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo
PubChem CID 81129
IUPAC Name 2-bromobenzaldehyde
SMILES BrC1=CC=CC=C1C=O

Specifications

Melting Point 18°C to 21°C
Density 1.597
Boiling Point 228°C to 230°C
Flash Point 95°C (203°F)
Odor Characteristic
Refractive Index 1.596
Quantity 25 g
Beilstein 636132
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol and benzene. Insoluble in water.
Formula Weight 185.03
Percent Purity 98%
Chemical Name or Material 2-Bromobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.