missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoanisole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.
Solubility
Soluble in ethanol and diethyl ether.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.
Solubility
Soluble in ethanol and diethyl ether.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | 2°C to 3°C |
Density | 1.511 |
Boiling Point | 220°C to 222°C |
Flash Point | 96°C (204°F) |
Assay Percent Range | 98% |
Refractive Index | 1.574 |
Quantity | 25 g |
Beilstein | 1859996 |
Solubility Information | Soluble in ethanol and diethyl ether. |
Formula Weight | 187.04 |
Show More |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'