missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanisole, 97%

Product Code. 10225890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225890 100 mL Glass bottle 100mL
10564111 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225890 Supplier Thermo Scientific Acros Supplier No. 170261000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 578-57-4 | C7H7BrO | 187.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-57-4
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00000064
InChI Key HTDQSWDEWGSAMN-UHFFFAOYSA-N
Synonym 2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
PubChem CID 11358
IUPAC Name 1-bromo-2-methoxybenzene
SMILES COC1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 2°C
Color Colorless to Yellow
Density 1.5000g/mL
Boiling Point 223°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OCH3
Refractive Index 1.5730 to 1.575
Quantity 100 mL
Beilstein 06, 197
Specific Gravity 1.5
Solubility Information Solubility in water: immiscible
Formula Weight 187.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromoanisole, 97%
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.