missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoadamantane, 98%

Product Code. 11409787
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11409787

Brand: Thermo Scientific Alfa Aesar B22760.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7314-85-4 | C10H15Br | 215.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7314-85-4
Molecular Formula C10H15Br
Molecular Weight (g/mol) 215.134
MDL Number MFCD00074745
InChI Key RCXJARRRXOPXBC-UHFFFAOYSA-N
Synonym 2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane
PubChem CID 522482
IUPAC Name 2-bromoadamantane
SMILES C1C2CC3CC1CC(C2)C3Br

Specifications

Melting Point 136°C to 141°C
Quantity 5 g
Beilstein 4655839
Formula Weight 215.14
Percent Purity 98%
Chemical Name or Material 2-Bromoadamantane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.