missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacrylic acid, 95%

Code produit. 15488789
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize'
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity unitSize
15488789 25 g 25g
11324504 1 g 1g
11334504 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 15488789 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L01194.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 10443-65-9 | C3H3BrO2 | 150.96 g/mol

2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoacrylic acid is used as a raw material on QSAR models to predict mutagenicity of acrylates. It is used as a pharmaceutical intermediate.

Solubility
It is miscible with water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10443-65-9
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.96
MDL Number MFCD00014333
InChI Key HMENQNSSJFLQOP-UHFFFAOYSA-N
Synonym 2-bromoacrylic acid, bromoacrylic acid, ccris 5478, .alpha.-bromoacrylic acid, 2-bromo-acrylic acid, alpha-bromo acrylic acid, 2-propenoic acid, bromo, acmc-1c0k4, 2-propenoic acid, 2-bromo
PubChem CID 82633
IUPAC Name 2-bromoprop-2-enoic acid
SMILES OC(=O)C(Br)=C

Spécification

Melting Point 65°C to 70°C
Quantity 25 g
UN Number UN3261
Beilstein 1740582
Solubility Information It is miscible with water.
Formula Weight 150.96
Percent Purity 95%
Chemical Name or Material 2-Bromoacrylic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.